N-Methyl-N-{[1-(3-methylphenyl)-1H-pyrazol-4-yl]methyl}amine

Supplier: Matrix Scientific

030934-500MG
101831-132EA 290.77 USD
101831-132
N-Methyl-N-{[1-(3-methylphenyl)-1H-pyrazol-4-yl]methyl}amine
N-Methyl-N-{[1-(3-methylphenyl)-1H-pyrazol-4-yl]methyl}amine

Hazard: Irritant

Formula: C₁₂H₁₅N₃
MW: 201.27 g/mol
MDL Number: MFCD08691448
CAS Number: 400756-89-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR