1-(2-Chloro-6-fluorobenzyl)-5-methyl-1H-pyrazol-3-amine

Supplier: Matrix Scientific

030898-500MG
101831-054EA 243.08 USD
101831-054
1-(2-Chloro-6-fluorobenzyl)-5-methyl-1H-pyrazol-3-amine
1-(2-Chloro-6-fluorobenzyl)-5-methyl-1H-pyrazol-3-amine

Hazard: Irritant

Formula: C₁₁H₁₁ClFN₃
MDL Number: MFCD04969987
CAS Number: 925200-31-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR