4-Allyl-5-[4-(diethylamino)phenyl]-4H-1,2,4-triazole-3-thiol

Supplier: Matrix Scientific

030634-500MG
101830-530EA 193.85 USD
101830-530
4-Allyl-5-[4-(diethylamino)phenyl]-4H-1,2,4-triazole-3-thiol
4-Allyl-5-[4-(diethylamino)phenyl]-4H-1,2,4-triazole-3-thiol

Hazard: Irritant

Formula: C₁₅H₂₀N₄S
MW: 288.42 g/mol
MDL Number: MFCD03900787
CAS Number: 669748-48-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR