4-Ethyl-5-(thien-2-ylmethyl)-4H-1,2,4-triazole-3-thiol

Supplier: Matrix Scientific

030492-500MG
101830-246EA 146.15 USD
101830-246
4-Ethyl-5-(thien-2-ylmethyl)-4H-1,2,4-triazole-3-thiol
4-Ethyl-5-(thien-2-ylmethyl)-4H-1,2,4-triazole-3-thiol

Hazard: Irritant

Formula: C₉H₁₁N₃S₂
MDL Number: MFCD03423486
CAS Number: 667435-85-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR