2-Pyridin-3-ylquinoline-4-carbohydrazide

Supplier: Matrix Scientific

029813-500MG
101828-908EA 290.77 USD
101828-908
2-Pyridin-3-ylquinoline-4-carbohydrazide
2-Pyridin-3-ylquinoline-4-carbohydrazide

Hazard: Irritant

Formula: C₁₅H₁₂N₄O
MDL Number: MFCD01993675
CAS Number: 5109-97-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR