2-Amino-N-(4-chlorophenyl)-5,6-dihydro-4H-cyclopenta[b]thiophene-3-carboxamide

Supplier: Matrix Scientific

029092-500MG
101827-458EA 193.85 USD
101827-458
2-Amino-N-(4-chlorophenyl)-5,6-dihydro-4H-cyclopenta[b]thiophene-3-carboxamide
2-Amino-N-(4-chlorophenyl)-5,6-dihydro-4H-cyclopenta[b]thiophene-3-carboxamide

Hazard: Irritant

Formula: C₁₄H₁₃ClN₂OS
MDL Number: MFCD01176829
CAS Number: 329066-94-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR