Dimethyl-5-amino-3-methylthiophene-2,4-dicarboxylate

Supplier: Matrix Scientific

028873-500MG
101827-020EA 64.62 USD
101827-020
Dimethyl-5-amino-3-methylthiophene-2,4-dicarboxylate
Dimethyl-5-amino-3-methylthiophene-2,4-dicarboxylate

Hazard: Irritant

Formula: C₉H₁₁NO₄S
MDL Number: MFCD00159553
CAS Number: 103765-33-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR