3-(4-Chloro-3-methyl-1H-pyrazol-1-yl)propanoic acid

Supplier: Matrix Scientific

027430-500MG
101824-178EA 387.69 USD
101824-178
3-(4-Chloro-3-methyl-1H-pyrazol-1-yl)propanoic acid
3-(4-Chloro-3-methyl-1H-pyrazol-1-yl)propanoic acid

Hazard: Irritant

Formula: C₇H₉ClN₂O₂
MW: 188.61 g/mol
MDL Number: MFCD03419612
CAS Number: 180741-32-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR