3-Aminoindan-1-one hydrochloride

Supplier: Matrix Scientific

Synonyms: 3-Amino-2,3-dihydro-1H-inden-1-one hydrochloride

027294-500MG
101823-906EA 495.38 USD
101823-906
3-Aminoindan-1-one hydrochloride
3-Aminoindan-1-one hydrochloride

Hazard: Irritant

Formula: C₉H₁₀ClNO
MW: 183.64 g/mol
MDL Number: MFCD11845790
CAS Number: 152605-34-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR