3-(Hexadecylamino)propanohydrazide

Supplier: Matrix Scientific

026883-500MG
101823-062EA 306.15 USD
101823-062
3-(Hexadecylamino)propanohydrazide
3-(Hexadecylamino)propanohydrazide

Hazard: Irritant

Formula: C₁₉H₄₁N₃O
MDL Number: MFCD10687294
CAS Number: 1040689-30-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR