3,6-Dibromo-2-picoline ≥98%

Supplier: Matrix Scientific

Synonyms: 3,6-Dibromo-2-methylpyridine, 2,5-Dibromo-6-methylpyridine

026296-5G
101822-018EA 23.08 USD
101822-018
3,6-Dibromo-2-picoline ≥98%
3,6-Dibromo-2-picoline

Hazard code(s): Irritant

Formula: C₆H₅Br₂N
MW: 250.92 g/mol
Melting Pt: 36…39 °C
MDL Number: MFCD06254589
CAS Number: 39919-65-8

Order Now

Specification Test Results

Melting point 36-39°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR