3-Amino-N-methylbenzamide

Supplier: Matrix Scientific

Synonyms: 3-Aminobenzoylmethylamide

025784-500MG
101820-946EA 1126.15 USD
101820-946
3-Amino-N-methylbenzamide
3-Amino-N-methylbenzamide

Hazard: Irritant

Formula: C₈H₁₀N₂O
MW: 150.18 g/mol
MDL Number: MFCD00070630
CAS Number: 25900-61-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR