N-(5-Amino-2-methoxyphenyl)-4-methoxybenzamide

Supplier: Matrix Scientific

025224-500MG
101819-826EA 306.15 USD
101819-826
N-(5-Amino-2-methoxyphenyl)-4-methoxybenzamide
N-(5-Amino-2-methoxyphenyl)-4-methoxybenzamide

Hazard: Irritant

Formula: C₁₅H₁₆N₂O₃
MW: 272.31 g/mol
MDL Number: MFCD09050199
CAS Number: 926190-58-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR