2-tert-Butyl-7-chloro-4-nitroindole

Supplier: Matrix Scientific

024965-250MG
101819-304EA 315.38 USD
101819-304
2-tert-Butyl-7-chloro-4-nitroindole
2-tert-Butyl-7-chloro-4-nitroindole

Hazard: Irritant

Formula: C₁₂H₁₃ClN₂O₂
MW: 252.7 g/mol
MDL Number: MFCD09743738
CAS Number: 1000018-53-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR