2-Amino-4-phenyl-5-phenylsulfonyl-1,3-thiazole

Supplier: Matrix Scientific

024929-500MG
101819-230EA 435.38 USD
101819-230
2-Amino-4-phenyl-5-phenylsulfonyl-1,3-thiazole
2-Amino-4-phenyl-5-phenylsulfonyl-1,3-thiazole

Hazard: Irritant

Formula: C₁₅H₁₂N₂O₂S₂
MW: 316.4 g/mol
MDL Number: MFCD03700392
CAS Number: 252679-72-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR