1-[4-(Ethylsulfonyl)-2-nitrophenyl]piperidine-4-carboxamide

Supplier: Matrix Scientific

024304-250MG
101817-838EA 315.38 USD
101817-838
1-[4-(Ethylsulfonyl)-2-nitrophenyl]piperidine-4-carboxamide
1-[4-(Ethylsulfonyl)-2-nitrophenyl]piperidine-4-carboxamide

Hazard: Irritant

Formula: C₁₄H₁₉N₃O₅S
MDL Number: MFCD08692377
CAS Number: 942474-30-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR