N-[4-(Hydrazinocarbonyl)phenyl]pentanamide

Supplier: Matrix Scientific

024011-500MG
101817-224EA 306.15 USD
101817-224
N-[4-(Hydrazinocarbonyl)phenyl]pentanamide
N-[4-(Hydrazinocarbonyl)phenyl]pentanamide

Hazard: Irritant

Formula: C₁₂H₁₇N₃O₂
MDL Number: MFCD06447091
CAS Number: 847468-37-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR