2-(2-Bromo-4-chlorophenoxy)acetohydrazide

Supplier: Matrix Scientific

024010-500MG
101817-222EA 306.15 USD
101817-222
2-(2-Bromo-4-chlorophenoxy)acetohydrazide
2-(2-Bromo-4-chlorophenoxy)acetohydrazide

Hazard: Irritant

Formula: C₈H₈BrClN₂O₂
MDL Number: MFCD00702269
CAS Number: 443908-11-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR