N-[4-(Hydrazinocarbonyl)phenyl]-3-methylbutanamide

Supplier: Matrix Scientific

024000-500MG
101817-202EA 306.15 USD
101817-202
N-[4-(Hydrazinocarbonyl)phenyl]-3-methylbutanamide
N-[4-(Hydrazinocarbonyl)phenyl]-3-methylbutanamide

Hazard: Irritant

Formula: C₁₂H₁₇N₃O₂
MDL Number: MFCD03306097
CAS Number: 496013-13-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR