N-(2,5-Dimethylphenyl)-4-hydrazino-4-oxobutanamide

Supplier: Matrix Scientific

023969-500MG
101817-140EA 306.15 USD
101817-140
N-(2,5-Dimethylphenyl)-4-hydrazino-4-oxobutanamide
N-(2,5-Dimethylphenyl)-4-hydrazino-4-oxobutanamide

Hazard: Irritant

Formula: C₁₂H₁₇N₃O₂
MDL Number: MFCD02105446
CAS Number: 315249-25-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR