2-(1-Benzyl-3-oxo-2-piperazinyl)acetic acid

Supplier: Matrix Scientific

023885-500MG
101816-968EA 306.15 USD
101816-968
2-(1-Benzyl-3-oxo-2-piperazinyl)acetic acid
2-(1-Benzyl-3-oxo-2-piperazinyl)acetic acid

Hazard: Irritant

Formula: C₁₃H₁₆N₂O₃
MDL Number: MFCD05666719
CAS Number: 693824-68-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR