Ethyl-2-(3-oxo-2-piperazinylidene)acetate

Supplier: Matrix Scientific

023856-500MG
101816-912EA 306.15 USD
101816-912
Ethyl-2-(3-oxo-2-piperazinylidene)acetate
Ethyl-2-(3-oxo-2-piperazinylidene)acetate

Hazard: Irritant

Formula: C₈H₁₂N₂O₃
MDL Number: MFCD00995938
CAS Number: 774-89-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR