2-[2-(4-Fluorophenoxy)ethyl]piperidine

Supplier: Matrix Scientific

023454-500MG
101816-036EA 315.38 USD
101816-036
2-[2-(4-Fluorophenoxy)ethyl]piperidine
2-[2-(4-Fluorophenoxy)ethyl]piperidine

Hazard: Irritant

Formula: C₁₃H₁₈FNO
MW: 223.29 g/mol
MDL Number: MFCD08687881
CAS Number: 475144-85-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR