3-[(4-Fluorobenzyl)oxy]pyrrolidine hydrochloride

Supplier: Matrix Scientific

023351-500MG
101815-832EA 186.15 USD
101815-832
3-[(4-Fluorobenzyl)oxy]pyrrolidine hydrochloride
3-[(4-Fluorobenzyl)oxy]pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₁H₁₅ClFNO
MW: 231.7 g/mol
MDL Number: MFCD10687224
CAS Number: 1185303-42-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR