4-(2-Methoxyethoxy)piperidine hydrochloride

Supplier: Matrix Scientific

023246-500MG
101815-626EA 315.38 USD
101815-626
4-(2-Methoxyethoxy)piperidine hydrochloride
4-(2-Methoxyethoxy)piperidine hydrochloride

Hazard: Irritant

Formula: C₈H₁₈ClNO₂
MDL Number: MFCD09879212
CAS Number: 550369-96-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR