8-(3-Piperidinyloxy)quinoline dihydrochloride

Supplier: Matrix Scientific

023230-500MG
101815-594EA 538.46 USD
101815-594
8-(3-Piperidinyloxy)quinoline dihydrochloride
8-(3-Piperidinyloxy)quinoline dihydrochloride

Hazard: Irritant

Formula: C₁₄H₁₈Cl₂N₂O
MW: 301.22 g/mol
MDL Number: MFCD12405044
CAS Number: 1185301-55-6

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR