4-Butoxy-2-(trifluoromethyl)aniline

Supplier: Matrix Scientific

022572-500MG
101814-300EA 315.38 USD
101814-300
4-Butoxy-2-(trifluoromethyl)aniline
4-Butoxy-2-(trifluoromethyl)aniline

Hazard: Irritant

Formula: C₁₁H₁₄F₃NO
MW: 233.24 g/mol
MDL Number: MFCD08687163
CAS Number: 121307-20-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR