4-(2-Methylphenoxy)-3-(trifluoromethyl)aniline

Supplier: Matrix Scientific

022514-500MG
101814-186EA 315.38 USD
101814-186
4-(2-Methylphenoxy)-3-(trifluoromethyl)aniline
4-(2-Methylphenoxy)-3-(trifluoromethyl)aniline

Hazard: Irritant

Formula: C₁₄H₁₂F₃NO
MW: 267.25 g/mol
MDL Number: MFCD08688336
CAS Number: 946775-76-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR