3-Chloro-4-(3-methoxypropoxy)phenylamine

Supplier: Matrix Scientific

022358-500MG
101813-876EA 315.38 USD
101813-876
3-Chloro-4-(3-methoxypropoxy)phenylamine
3-Chloro-4-(3-methoxypropoxy)phenylamine

Hazard: Irritant

Formula: C₁₀H₁₄ClNO₂
MDL Number: MFCD08687370
CAS Number: 946775-28-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR