2-(2-Methylphenoxy)-5-(trifluoromethyl)aniline

Supplier: Matrix Scientific

022052-500MG
101813-266EA 315.38 USD
101813-266
2-(2-Methylphenoxy)-5-(trifluoromethyl)aniline
2-(2-Methylphenoxy)-5-(trifluoromethyl)aniline

Hazard: Irritant

Formula: C₁₄H₁₂F₃NO
MW: 267.25 g/mol
MDL Number: MFCD08688333
CAS Number: 401-20-7

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR