5-Chloro-2-(3-methoxypropoxy)phenylamine

Supplier: Matrix Scientific

021967-500MG
101813-096EA 315.38 USD
101813-096
5-Chloro-2-(3-methoxypropoxy)phenylamine
5-Chloro-2-(3-methoxypropoxy)phenylamine

Hazard: Irritant

Formula: C₁₀H₁₄ClNO₂
MDL Number: MFCD08687371
CAS Number: 946682-50-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR