3-Chloro-2-(2-methoxyphenoxy)phenylamine

Supplier: Matrix Scientific

021896-500MG
101812-954EA 315.38 USD
101812-954
3-Chloro-2-(2-methoxyphenoxy)phenylamine
3-Chloro-2-(2-methoxyphenoxy)phenylamine

Hazard: Irritant

Formula: C₁₃H₁₂ClNO₂
MDL Number: MFCD08687363
CAS Number: 946714-63-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR