4-(2,5-Dimethylphenyl)piperidine hydrochloride

Supplier: Matrix Scientific

021729-500MG
101812-632EA 315.38 USD
101812-632
4-(2,5-Dimethylphenyl)piperidine hydrochloride
4-(2,5-Dimethylphenyl)piperidine hydrochloride

Hazard: Irritant

Formula: C₁₃H₂₀ClN
MDL Number: MFCD11100470
CAS Number: 1004618-86-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR