2-Methyl-5-trifluoromethyl-2H-pyrazole-3-carboxylic acid

Supplier: Matrix Scientific

Synonyms: 1-Methyl-3-(trifluoromethyl)-1H-pyrazole-5-carboxylic acid

021545-500MG
101812-286EA 307.69 USD
101812-286
2-Methyl-5-trifluoromethyl-2H-pyrazole-3-carboxylic acid
2-Methyl-5-trifluoromethyl-2H-pyrazole-3-carboxylic acid

Hazard: Irritant

Formula: C₆H₅F₃N₂O₂
MW: 194.11 g/mol
MDL Number: MFCD02090853
CAS Number: 128694-63-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR