C-(1-Ethylpyrrolidin-3-yl)methylamine

Supplier: Matrix Scientific

021487-500MG
101812-176EA 261.54 USD
101812-176
C-(1-Ethylpyrrolidin-3-yl)methylamine
C-(1-Ethylpyrrolidin-3-yl)methylamine

Hazard: Irritant

Formula: C₇H₁₆N₂
MDL Number: MFCD05864572
CAS Number: 51388-01-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR