4-(3-Fluorobenzyloxy)phenylamine

Supplier: Matrix Scientific

Synonyms: 4-((3-Fluorobenzyl)oxy)aniline

020400-500MG
101809-960EA 280 USD
101809-960
4-(3-Fluorobenzyloxy)phenylamine
4-(3-Fluorobenzyloxy)phenylamine

Hazard: Irritant

Formula: C₁₃H₁₂FNO
MW: 217.25 g/mol
MDL Number: MFCD08056104
CAS Number: 202197-25-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR