1-Ethyl-1H-pyrazole-4-sulfonyl chloride

Supplier: Matrix Scientific

019863-500MG
101808-938EA 64.62 USD
101808-938
1-Ethyl-1H-pyrazole-4-sulfonyl chloride
1-Ethyl-1H-pyrazole-4-sulfonyl chloride

Hazard: Irritant

Formula: C₅H₇ClN₂O₂S
MDL Number: MFCD04968670
CAS Number: 957514-21-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR