2-(2-Methyl-1H-indol-1-yl)acetic acid

Supplier: Matrix Scientific

018450-500MG
101806-224EA 280 USD
101806-224
2-(2-Methyl-1H-indol-1-yl)acetic acid
2-(2-Methyl-1H-indol-1-yl)acetic acid

Hazard: Irritant

Formula: C₁₁H₁₁NO₂
CAS Number: 86704-55-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR