6,7-Diiodobenzo(1,4)dioxan ≥98%

Supplier: Matrix Scientific

016654-500MG
101805-148EA 213.85 USD
101805-148
6,7-Diiodobenzo(1,4)dioxan ≥98%
6,7-Diiodobenzo(1,4)dioxan

Hazard: Irritant

Formula: C₈H₆I₂O₂
MDL Number: MFCD06656536
CAS Number: 155303-91-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR