N-Methyl-1-[3-(1H-pyrazol-1-yl)phenyl]methanamine dihydrochloride

Supplier: Matrix Scientific

016270-500MG
101804-416EA 283.08 USD
101804-416
N-Methyl-1-[3-(1H-pyrazol-1-yl)phenyl]methanamine dihydrochloride
N-Methyl-1-[3-(1H-pyrazol-1-yl)phenyl]methanamine dihydrochloride

Hazard: Irritant

Formula: C₁₁H₁₅Cl₂N₃
MDL Number: MFCD06801104
CAS Number: 1185294-98-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR