3-Thiocarbamoylpyrrolidine-1-carboxylic acid tert-butyl ester

Supplier: Matrix Scientific

Synonyms: tert-Butyl 3-carbamothioylpyrrolidine-1-carboxylate

016049-500MG
101803-972EA 249.23 USD
101803-972
3-Thiocarbamoylpyrrolidine-1-carboxylic acid tert-butyl ester
3-Thiocarbamoylpyrrolidine-1-carboxylic acid tert-butyl ester

Hazard: Irritant

Formula: C₁₀H₁₈N₂O₂S
MW: 230.33 g/mol
Melting Pt: 131…133 °C
MDL Number: MFCD03791259
CAS Number: 122684-35-9

Order Now

Specification Test Results

Melting point 139-141°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR