2-(4-Fluorophenoxy)-N-methyl-1-ethanamine

Supplier: Matrix Scientific

013768-500MG
101803-686EA 278.46 USD
101803-686
2-(4-Fluorophenoxy)-N-methyl-1-ethanamine
2-(4-Fluorophenoxy)-N-methyl-1-ethanamine

Hazard: Irritant

Formula: C₉H₁₂FNO
MDL Number: MFCD06247667
CAS Number: 694431-80-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR