Octahydro-2H-pyrido[1,2-a]pyrazine

Supplier: Matrix Scientific

Synonyms: 1,4-Diazabicyclo[4.4.0]decane

011636-1G
101801-030EA 643.08 USD
101801-030
Octahydro-2H-pyrido[1,2-a]pyrazine
Octahydro-2H-pyrido[1,2-a]pyrazine

Hazard: Irritant

Formula: C₈H₁₆N₂
MW: 140.23 g/mol
Boiling Pt: 84 °C (12 Torr)
MDL Number: MFCD01098257
CAS Number: 4430-75-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR