4-[(4-Bromophenyl)sulfonyl]piperidine hydrochloride

Supplier: Matrix Scientific

011078-1G
101799-790EA 283.08 USD
101799-790
4-[(4-Bromophenyl)sulfonyl]piperidine hydrochloride
4-[(4-Bromophenyl)sulfonyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₁H₁₅BrClNO₂S
MDL Number: MFCD04116083
CAS Number: 226400-32-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR