2-(Benzylaminomethyl)phenol hydrochloride

Supplier: Matrix Scientific

011055-500MG
101799-740EA 267.69 USD
101799-740
2-(Benzylaminomethyl)phenol hydrochloride
2-(Benzylaminomethyl)phenol hydrochloride

Hazard: Irritant

Formula: C₁₄H₁₆ClNO
Melting Pt: 190…191 °C
MDL Number: MFCD00663262
CAS Number: 73057-58-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR