1-(Methylsulfonyl)-4-piperidinecarboxylic acid

Supplier: Matrix Scientific

Synonyms: 1-(Methylsulphonyl)piperidine-4-carboxylic acid

010890-500MG
101799-416EA 138.46 USD
101799-416
1-(Methylsulfonyl)-4-piperidinecarboxylic acid
1-(Methylsulphonyl)-4-piperidinecarboxylic acid

Hazard: Irritant

Formula: C₇H₁₃NO₄S
MW: 207.2500 g/mol
Melting Pt: 161-163°C
MDL Number: MFCD03821886
CAS Number: 280772-00-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR