1H-(N-Benzyl)-4-iodopyrazole ≥98%

Supplier: Matrix Scientific

Synonyms: 1-Benzyl-4-iodo-1H-pyrazole

009754-1G
101797-174EA 150.77 USD
101797-174
1H-(N-Benzyl)-4-iodopyrazole ≥98%
1H-(N-Benzyl)-4-iodopyrazole

Hazard: Irritant

Formula: C₁₀H₉IN₂
MW: 284.1 g/mol
MDL Number: MFCD04038745
CAS Number: 50877-42-4

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR