N-(2-Aminoethyl)-N-[4-(methylsulfonyl)-2-nitrophenyl]amine

Supplier: Matrix Scientific

009367-1G
101796-946EA 138.46 USD
101796-946
N-(2-Aminoethyl)-N-[4-(methylsulfonyl)-2-nitrophenyl]amine
N-(2-Aminoethyl)-N-[4-(methylsulfonyl)-2-nitrophenyl]amine

Hazard: Irritant

Formula: C₉H₁₃N₃O₄S
MW: 259.29 g/mol
Melting Pt: 144…145 °C
MDL Number: MFCD04037966
CAS Number: 849035-93-4

Order Now

Specification Test Results

Melting point 144-145°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR