6-(Chloromethyl)-2,3-dihydro-1H,5H-pyrido[3,2,1-ij]quinolin-5-one

Supplier: Matrix Scientific

009287-500MG
101796-788EA 184.62 USD
101796-788
6-(Chloromethyl)-2,3-dihydro-1H,5H-pyrido[3,2,1-ij]quinolin-5-one
6-(Chloromethyl)-2,3-dihydro-1H,5H-pyrido[3,2,1-ij]quinolin-5-one

Hazard: Irritant

Formula: C₁₃H₁₂ClNO
MW: 233.7 g/mol
MDL Number: MFCD04037905
CAS Number: 849021-07-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR