1-Fluoropropane ≥98%

Supplier: Matrix Scientific

Synonyms: Propyl fluoride

006055-10G
101789-984EA 484.62 USD
101789-984
1-Fluoropropane ≥98%
1-Fluoropropane

Hazard: Flammable Gas

Formula: C₃H₇F
MW: 62.09 g/mol
Boiling Pt: -2.5°C
Melting Pt: –159 °C
MDL Number: MFCD01320734
CAS Number: 460-13-9

Order Now

Specification Test Results

Boiling point -2.5°C
Melting point -159°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR